Z215511
Rigid PTFE sleeve
joint: ST/NS 29/32
Sinonimo/i:
ptfe sleeves
About This Item
Prodotti consigliati
Giunto
joint: ST/NS29/32
Produttore/marchio commerciale
Cowie
Descrizione generale
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.