Documenti
Z171697
Ace double-jacketed coiled condenser
jacket L 300 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Giunto
joint ST/NS24/40
Produttore/marchio commerciale
Ace Glass 595514
Lunghezza camicia
300 mm
Categorie correlate
Descrizione generale
Extra cooling capacity for volatile materials. Compact design retards diffusion.