Documenti
Z107255
Sigma-Aldrich
Aldrich® round-bottom storage flask with stopcock-equipped septum-inlet
with 2 mm glass stopcock, capacity 50 mL, joint: ST/NS 14/20
Sinonimo/i:
bottom flask, flask round, round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
round bottom flask
two-neck
Giunto
joint: ST/NS14/20
Sterilità
non-sterile
Caratteristiche
closure type PTFE stopcock
Confezionamento
pack of 1
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
50 mL
Descrizione generale
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa Section).
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC