Documenti
Z100331
Sigma-Aldrich
Aldrich® single-neck round-bottom flask with Clear-Seal joint
capacity 50 mL
Sinonimo/i:
bottom flask, flask round, round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
round bottom flask
single-neck
Giunto
female joint ST/NS19/22
Sterilità
non-sterile
Caratteristiche
closure type Standard Taper joint
Confezionamento
pack of 1
Produttore/marchio commerciale
Sigma-Aldrich
Capacità
50 mL
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC