Documenti
WHA68822516
Filtri da siringa Whatman® GD/X
GF/A membrane, pore size 1.6 μm, diam. 25 mm, non-sterile, pk of 150 ea
Sinonimo/i:
Whatman filter, Z745413, syringe filter, syringe filters
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Materiali
glass microfiber membrane
polypropylene housing
Sterilità
non-sterile
Caratteristiche
glass fiber prefilter
Confezionamento
pk of 150 ea
Produttore/marchio commerciale
Whatman 6882-2516
Whatman Article No. 28420531 (US reference)
Diametro
25 mm
Volume
≤25 mL
Dimensione pori
1.6 μm
Raccordi
female Luer inlet (lock)
male Luer outlet
Descrizione generale
Non-sterile syringe filters with a glass membrane, suitable for hard-to-filter material with high solids. Whatman GD/X Syringe Filters are high-viscosity filters that incorporate a prefilter to enable filtration of viscous, dense or otherwise hard-to-filter material with high solids content as in environmental and food analysis samples. The prefilter contains a high-efficiency borosilicate glass filtration stack that will retain fine particles down to 0.7 μm, allowing difficult sample filtration with less hand force. It reduces the need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters. The filters can be autoclaved.
Caratteristiche e vantaggi
  • Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
  • Polyethersulfone (PES) for serum, plasma and tissue culture
  • Polyvinylidene difluoride (PVDF) for HPLC
  • Cellulose acetate for biological solutions
  • Borosilicate glass microfiber
Compatibilità
Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva