Documenti
SYNS461230
Synthware straight bore glass stopcock plug
bore size 2 mm, plug size 12/30
Sinonimo/i:
Synthware S461230
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware S461230
Dimensioni foro
2 mm
Categorie correlate
Descrizione generale
To be used with constant pressure retention kit (S69)
Note legali
Synthware is a trademark of Kemtech America Inc.