Documenti
SYNF621960
Synthware pressure equalizing addition funnel
60 mL, joint: ST/NS 19/20, stopcock: PTFE
Sinonimo/i:
Synthware F621960, dropping funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Giunto
joint: ST/NS19/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F621960
Capacità
60 mL
60 mL
Descrizione generale
With 1:5 PTFE stopcock
Note legali
Synthware is a trademark of Kemtech America Inc.