Scegli un formato
| A voi/SKU | Disponibilità | Prezzo |
|---|---|---|
1 ea | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 155,00 € 139,50 € |
Informazioni su questo articolo
139,50 €
Prezzo di listino155,00 €Risparmia il 10%material
borosilicate glass flask, four-neck , round bottom
joint
center joint: ST/NS24/40, side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F214500
capacity
500 mL
General description
Legal Information
1 of 1
Questo articolo | |||
|---|---|---|---|
| description capacity 500 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description capacity 500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40 | description capacity 500 mL, center joint: ST/NS 29/42, side joint: ST/NS 29/42 | description capacity 1,000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 |
| capacity 500 mL | capacity 500 mL | capacity 500 mL | capacity 1,000 mL |
| manufacturer/tradename Synthware F214500 | manufacturer/tradename Synthware F212500 | manufacturer/tradename Synthware F218500 | manufacturer/tradename Synthware F214000 |
| joint side joint: ST/NS24/40, center joint: ST/NS24/40 | joint center joint: ST/NS29/42, side joint: ST/NS24/40 | joint side joint: ST/NS29/42, center joint: ST/NS29/42 | joint center joint: ST/NS24/40, side joint: ST/NS24/40 |
| material round bottom , four-neck , borosilicate glass flask | material four-neck , borosilicate glass flask, round bottom | material four-neck , borosilicate glass flask, round bottom | material four-neck , borosilicate glass flask, round bottom |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ four-neck round-bottom flask with vertical side necks
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?