SYNA971922
Synthware™ distilling adapter with compression cap
joint: ST/NS 19/22, side arm angle: 75 deg
Sinonimo/i:
Synthware A971922, adapter, distillation adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS19/22
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A971922
Categorie correlate
Descrizione generale
With compression cap (RC009) and Viton® O-ring for forming a vacuum tight seal of plain thermometer.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)