Documenti
SYNA872440
Synthware 75 degree angled drying tube
joint: ST/NS 24/40
Sinonimo/i:
Synthware A872440, glass drying tube, Drying tube 75° bent
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Materiali
colorless tube
glass tube
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A872440
Descrizione generale
Angled at 75° to avoid desiccant contact with reactions.
Note legali
Synthware is a trademark of Kemtech America Inc.