MT51344622
Mettler Toledo InLab® OptiOx dissolved oxygen sensor
cable L 5 m
About This Item
Codice UNSPSC:
39121436
NACRES:
NB.79
Descrizione
0 - 500%, 0 - 50 mg/L
mini LTW connector
Produttore/marchio commerciale
Mettler-Toledo 51344622
Lungh. cavo
5 m
Descrizione generale
The InLab® OptiOx-5m optical dissolved oxygen sensor is based on the reliable RDO technology. Thanks to RDO (Rugged Dissolved Oxygen), measuring dissolved oxygen is easier than ever before:
In combination with ISM® (intelligent sensor management) you will find the high accuracy and reproducibility that you have come to trust with METTLER TOLEDO products.
The InLab® OptiOx sensors can be used in combination with a SevenExcellence benchtop meter or the SevenGo 9 or SevenGo Duo 98 portable meters.
Features and Benefits
The InLab® OptiOx-5m for determining dissolved oxygen is based on the reliable optical RDO (Rugged Dissolved Oxygen) technology.
RDO is fast, precise and produces extremely reproducible results.
Thanks to the robust design and the customized accessories, the InLab® OptiOx-5 m is a true all rounder. As a result, it′s ideally equipped for use in harsh, field based environments (when fitted with the OptiOx protective guard) and in the laboratory as well.
- Stable results and rapid response time
- Immediately ready for use
- Extremely easy handling, zero maintenance: save time!
- Suitable for an extremely wide range of applications
In combination with ISM® (intelligent sensor management) you will find the high accuracy and reproducibility that you have come to trust with METTLER TOLEDO products.
The InLab® OptiOx sensors can be used in combination with a SevenExcellence benchtop meter or the SevenGo 9 or SevenGo Duo 98 portable meters.
Features and Benefits
The InLab® OptiOx-5m for determining dissolved oxygen is based on the reliable optical RDO (Rugged Dissolved Oxygen) technology.
RDO is fast, precise and produces extremely reproducible results.
Thanks to the robust design and the customized accessories, the InLab® OptiOx-5 m is a true all rounder. As a result, it′s ideally equipped for use in harsh, field based environments (when fitted with the OptiOx protective guard) and in the laboratory as well.
Note legali
ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)