Documenti
GF89954779
Platinum-iridium alloy, Pt90Ir10
foil, 25 x 25mm, 0.05mm thickness, as rolled, light tight (lt)
Sinonimo/i:
PT020240
About This Item
Codice UNSPSC:
11101711
NACRES:
NA.23
Descrizione
light tight (lt)
Forma fisica
foil (as rolled)
Produttore/marchio commerciale
Goodfellow 899-547-79
Misura × spessore
25 mm × 25 mm × 0.05 mm