Documenti fondamentali
CLS70320250
Pyrex® VISTA round-bottom boiling flask
capacity 250 mL
Sinonimo/i:
boiling flask, bottom flask, round bottomed flask, Corning® 70320
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
round bottom flask
short
Giunto
joint ST/NS24/40
Sterilità
non-sterile
Caratteristiche
closure type Standard taper joint
Confezionamento
pack of 2
case of 4
Produttore/marchio commerciale
Corning 70320-250
Capacità
250 mL
Dimens. collo
(1)
Descrizione generale
These round bottom boiling flasks have full length outer joints, but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)