Documenti
CLS53201L
Pyrex® heavy-wall graduated filtering flask
capacity 1,000
Sinonimo/i:
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
flat bottom flask
narrow-mouth
Giunto
joint stopper8
Descrizione
without hose connection
Sterilità
non-sterile
Caratteristiche
closure type Rubber stopper
Confezionamento
case of 12
pack of 6
Produttore/marchio commerciale
Corning 5320-1L
Campo di misura del volume
300-1,000 mL
Capacità
1,000
Descrizione generale
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.