Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|---|---|
10 ea | Per informazioni sulla disponibilità, contatta il Servizio Clienti. | 132,00 € |
Informazioni su questo articolo
132,00 €
material
polystyrene , transparent wells
sterility
sterile
feature
lid, single packed
manufacturer/tradename
BRAND 782890
packaging
pack of 10 ea, pkg of Individually wrapped, with lid
size
24 well
suitability
suitable for (bacteriological assays)
General description
Features and Benefits
- Optimal cell growth thanks to cellGrade™ plus surface
- Side well access for easier pipetting and removing cover slips
- Perfect positioning of the BRANDplates® insert
Legal Information
1 of 1
Questo articolo | ||
|---|---|---|
| description 24-well with lid cellGrade™ plus | description 24-well with lid cellGrade™ S | description 6-well with lid cellGrade™ plus |
| size 24 well | size 24 well | size 6 well |
| suitability suitable for (bacteriological assays) | suitability suitable for (bacteriological assays) | suitability suitable for (bacteriological assays) |
| material polystyrene , transparent wells | material transparent wells, polystyrene | material transparent wells |
| feature single packed, lid | feature lid, single packed | feature lid, single packed |
| sterility sterile | sterility sterile | sterility sterile |
| packaging pack of 10 ea, pkg of Individually wrapped, with lid | packaging pack of 10 ea, pkg of Individually wrapped, with lid | packaging pkg of Individually wrapped, with lid, pack of 1 ea |
Still not finding the right product?
Explore all of our products under BRAND® BRANDplates® for inserts
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?