Documenti
566152
Sigma-Aldrich
Bis(pentafluorophenyl)((trifluoromethanesulfonyl)oxy)borane
About This Item
Formula condensata:
(C6F5)2BOSO2CF3
Numero CAS:
Peso molecolare:
493.99
Codice UNSPSC:
12352103