ZP-1G-PUR
Recombinant Mouse IgG B1
bottle of 1 g
Synonym(s):
Antibody Blocker, HAMA, Passive Antibody Blocker, Purified Mouse IgG, Recombinant Mouse IgG
About This Item
Recommended Products
biological source
mouse
antibody form
purified immunoglobulin
material
PETG bottle
clone
recombinant monoclonal
description
Phosphate Buffer Saline
Assay
≥90% (HPLC)
form
liquid
packaging
bottle of 1 g
concentration
~5 mg/mL
technique(s)
ELISA: suitable
immunoblotting: suitable
lateral flow assay: suitable
pH
7.2
isotype
IgG
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
The recombinant mouse IgG is offered in two unique blends providing different formulations to select from and optimize for best performance results within a specific immunoassay. The formulas are designed to deliver equivalent or better blocking efficiency compared to serum-derived mouse IgG. This B1 recombinant mouse IgG was developed as a blocking reagent to passively bind human anti-mouse antibodies present in certain serum/plasma samples and reduce non-specific binding in an immunoassay system using capture and detection antibodies.
Features and Benefits
- Sustainably sourced requiring no animal involvement.
- Batch reproducibility with clonal-derived, cultured supernatant instead of pooled serum.
- Security of scalable supply with on-time delivery.
- Tightly controlled product manufacturing for optimal performance.
- Simplified import regulations and restrictions.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Recombinant mouse IgG antibody offers controlled, non-animal derived blocking solution for immunoassay manufacturers.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service