Skip to Content
Merck
All Photos(2)

Documents

850101P

Avanti

13:0 Lyso PI

1-tridecanoyl-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt), powder

Synonym(s):

PI(13:0/0:0); 110716

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H46NO12P
CAS Number:
Molecular Weight:
547.57
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (LPI; may contain up to 10% of the 2-LPI isomer, TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850101P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850101P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850101P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(O)COC(CCCCCCCCCCCC)=O.[NH4+]

General description

Lysophosphatidylinositol (LPI), produced by phospholipase A2 is a bioactive lipid.

Application

13:0 Lyso PI (1-tridecanoyl-2-hydroxy-sn-glycero-3-phospho-(1′-myo-inositol) may be used as a nonendogenous phospholipid (PL) standard for the quantitative analysis of PLs. It may also be used as a standard for tandem mass spectrometry analysis of lipid species in human plasma.

Biochem/physiol Actions

Increased circulating levels of L-alpha-lysophosphatidylinositol (LPI) are associated with cancer and LPI is a potent ligand for the G-protein-coupled receptor GPR55.
Lysophosphatidylinositol (LPI) is expected to play a vital role in various diseases. Ca2+ channels at the plasma membrane and mitochondrial membrane are controlled by LPI. It can also stimulate various signaling pathways to control several cellular processes. LPI can block the migration of cells.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850101P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850101P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Piñeiro et al.
Biochimica et biophysica acta, 1821(4), 694-705 (2012-01-31)
Lysophosphatidylinositol (LPI) is a bioactive lipid generated by phospholipase A2 which is believed to play an important role in several diseases. Indeed LPI can affect various functions such as cell growth, differentiation and motility, in a number of cell-types, including
Shuang Song et al.
Food chemistry, 240, 1171-1178 (2017-09-28)
Nuts, a class of fatty natural food, are associated with a series of health benefit. Lipid, which is the most abundant nutrient in nuts, could be one of the major contributors to many beneficial effects. In previous studies, only the
Kevin Huynh et al.
Cell chemical biology, 26(1), 71-84 (2018-11-13)
High-throughput targeted lipid profiling with liquid chromatography-mass spectrometry (LC-MS) has been used extensively to identify associations between plasma lipid species and disease states. Such methods, used to characterize larger clinical cohorts, often suffer from an inability to differentiate isomeric forms
David B Iaea et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158609-158609 (2020-01-10)
STARD4, a member of the evolutionarily conserved START gene family, is a soluble sterol transport protein implicated in cholesterol sensing and maintenance of cellular homeostasis. STARD4 is widely expressed and has been shown to transfer sterol between liposomes as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service