Skip to Content
Merck
All Photos(1)

Documents

SAB1406282

Sigma-Aldrich

Anti-PPP1CB antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC3672, PP-1B, PPP1CD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1CB(5500)

Related Categories

General description

Protein phosphatase 1 catalytic subunit β (PPP1CB), is encoded by the gene mapped to human chromosome 2p23.2. The protein is expressed in spines, dendrites, axon terminals, axons, and glia in the prefrontal cortex, but at higher levels in dendrites. PPP1CB is characterized with a calcineurin-like phosphoesterase domain, a metallophosphatase domain and a serine/threonine protein phosphatase domain.
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed. (provided by RefSeq)

Immunogen

PPP1CB (NP_002700.1, 1 a.a. ~ 327 a.a) full-length human protein.

Sequence
MADGELNVDSLITRLLEVRGCRPGKIVQMTEAEVRGLCIKSREIFLSQPILLELEAPLKICGDIHGQYTDLLRLFEYGGFPPEANYLFLGDYVDRGKQSLETICLLLAYKIKYPENFFLLRGNHECASINRIYGFYDECKRRFNIKLWKTFTDCFNCLPIAAIVDEKIFCCHGGLSPDLQSMEQIRRIMRPTDVPDTGLLCDLLWSDPDKDVQGWGENDRGVSFTFGADVVSKFLNRHDLDLICRAHQVVEDGYEFFAKRQLVTLFSAPNYCGEFDNAGGMMSVDETLMCSFQILKPSEKKAKYQYGGLNSGRPVTPPRTANPPKKR

Biochem/physiol Actions

Protein phosphatase 1 catalytic subunit β (PPP1CB) plays a vital role in various cellular processes, such as cell adhesion, cell cycle, small GTPase-mediated signal transduction, protein dephosphorylation, negative regulation of transforming growth factor, regulation of glycogen catabolic process and striated muscle tissue development. In brain, PPP1CB regulates synaptic plasticity. Mutation in the gene causes intellectual disability (ID), congenital heart disease. In addition, variation in the gene expression is also associated with the development of features resembling noonan syndrome with loose anagen hair (NS-LAH).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Rasopathy Caused by Recurrent De Novo Missense Mutations In PPP1CB Closely Resembles Noonan Syndrome with Loose Anagen Hair
Gripp KW, et al.
American Journal of Medical Genetics. Part A, 170(9), 2237-2247 (2016)
De Novo Missense Variants in PPP1CB Are Associated with Intellectual Disabilities and Congenital Heart Disease
Ma L, et al.
Human Genetics, 135(12), 1399-1409 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service