Skip to Content
Merck
All Photos(2)

Documents

SAB56000308

Sigma-Aldrich

p16 INK4a Rabbit Monoclonal Antibody

clone RM409, affinity purified immunoglobulin

Synonym(s):

Cyclin-dependent kinase 4 inhibitor A, Cyclin-dependent kinase inhibitor 2A, p16-INK4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

affinity purified immunoglobulin

Quality Level

antibody product type

primary antibodies

clone

RM409, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:200
western blot: 1:1000-1:2000

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

Specificity

This antibody reacts to human p16.

Immunogen

A peptide corresponding to C-terminus of human p16

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service