Skip to Content
Merck
All Photos(1)

Key Documents

C5599

Sigma-Aldrich

Z-Ile-Glu-Thr-Asp 7-amido-4-trifluoromethylcoumarin

≥95% (HPLC)

Synonym(s):

Cbz-IETD-AFC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H42F3N5O13
CAS Number:
Molecular Weight:
821.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)OCc1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc2ccc3c(OC(=O)C=C3C(F)(F)F)c2

InChI

1S/C37H42F3N5O13/c1-4-18(2)30(45-36(56)57-17-20-8-6-5-7-9-20)34(54)42-24(12-13-27(47)48)32(52)44-31(19(3)46)35(55)43-25(16-28(49)50)33(53)41-21-10-11-22-23(37(38,39)40)15-29(51)58-26(22)14-21/h5-11,14-15,18-19,24-25,30-31,46H,4,12-13,16-17H2,1-3H3,(H,41,53)(H,42,54)(H,43,55)(H,44,52)(H,45,56)(H,47,48)(H,49,50)/t18-,19+,24-,25-,30-,31-/m0/s1

InChI key

RCIFCAJYIPUCJW-QXPVPWKFSA-N

Amino Acid Sequence

Z-Ile-Glu-Thr-Asp-AFC

Application

Fluorogenic substrate for caspase 8 and granzyme B.

Biochem/physiol Actions

IETD is the sequence of amino acid residues 172-175 of procaspase 3, which is cleaved during apoptosis to produce the p12 subunit and a p20 peptide that is further cleaved to form the p17 subunit of mature caspase 3.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Han et al.
The Journal of biological chemistry, 272(20), 13432-13436 (1997-05-16)
The apoptotic cysteine protease, caspase-3, is expressed in cells as an inactive 32-kDa precursor from which 17 kDa (p17) and 12 kDa (p12) subunits of the mature caspase-3 are proteolytically generated during apoptosis. Two amino acid sequences, ESMD downward arrowS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service