Z802808
Hellma® TrayCell™ cap
pathlength 1 (factor 10) mm ± 0.02 mm, chamber volume 3-5 μL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
Recommended Products
manufacturer/tradename
Hellma (665.703)
technique(s)
UV/Vis spectroscopy: suitable
photometry: suitable
chamber volume
3-5 μL
pathlength
1 (factor 10) mm ± 0.02 mm
Looking for similar products? Visit Product Comparison Guide
General description
Cap made of stainless steel with a mirror made of Quartz Suprasil® with an aluminum mirror layer.
factor = dilution factor compared to a standard cell with a path length of 10 mm
factor = dilution factor compared to a standard cell with a path length of 10 mm
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
TrayCell is a trademark of Hellma GmbH
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service