Skip to Content
Merck
All Photos(1)

Key Documents

00630

Sigma-Aldrich

Diethyl 1,3-acetonedicarboxylate

technical, ≥90% (GC)

Synonym(s):

Diethyl 3-oxoglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(CH2COOC2H5)2
CAS Number:
Molecular Weight:
202.20
Beilstein:
640146
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

refractive index

n20/D 1.440 (lit.)

bp

250 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)CC(=O)OCC

InChI

1S/C9H14O5/c1-3-13-8(11)5-7(10)6-9(12)14-4-2/h3-6H2,1-2H3

InChI key

ZSANYRMTSBBUCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service