Skip to Content
Merck
All Photos(2)

Documents

1.09191

Millipore

MAS-100 NT®

Portable Microbial Air Sampler for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms.

Synonym(s):

microbial air monitoring systems, Portable air sampler

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

aluminum hardware

Quality Level

Agency

EN 17141
ISO 14698-1
USP 797

product line

MAS-100®

feature

portable

packaging

pkg of 1 kit

manufacturer/tradename

MAS-100 NT®

technique(s)

active air sampling: suitable

H × diam.

25 cm × 11 cm

weight

2.3 kg (including perforated lid)

pore size

0.6 mm pore size

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with 90 mm Petri dishes

General description

The MAS-100 NT® microbial air monitoring systems are high performance, portable, battery operated instrument which are based on the principle of the Andersen air sampler. These compact yet sophisticated devices are the preferred choice for those demanding the highest quality in microbial air monitoring. A specified amount of air is aspirated through a 300-hole x 0.6 mm perforated lid and impacted onto the surface of growth media in standard 90-100 mm Petri dishes. Also 55-60 mm contact plates can be used with specified adapters and perforated lids. Microorganisms are impacted onto agar and, after an appropriate incubation period, the colonies are counted. The system measures the airflow and regulates it to a constant value of 100 standard liters/min. A Standard Liter Per Minute (SLPM) is adjusted to a specific environmental temperature and pressure, this ensure the amount of air sampled is always the same and independent of environmental conditions. These systems have the highest airflow accuracy of ± 2.5%, compared to others that can be as high as ±10%. The integrated flow sensor and high precision of perforated lids allows the user to freely interchange the perforated lids without affecting the accuracy or the calibration of the unit. The instruments are independently validated according to ISO 14698-1 with very good physical efficiency providing a D50 value below 1μm and a high biological efficiency result. The MAS-100 NT® is powered by a Lithium ion rechargeable battery with an intelligent charging program that assures long battery life without routine discharging.

Application

The MAS-100 NT® air sampler is used for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications. Additionally, a regulation for compounding pharmacies, USP797, that requires active air sampling in the level 5 and above areas.

Components

Includes hard carrying case, battery, 300-hole perforated lid, dust cover, main charger, USB cable, 3 mm allen key, and USB stick with all documentation

Other Notes

Now available: The new Safe Lid for the MAS-100 NT® air sampler

Safe Lid is a disposable, ready-to use alternative to the autoclavable standard perforated lid made from aluminum. It is made of 100% recyclable Acrylonitrile Butadiene Styrene (ABS) and comes in 30 individually double-packed and sterilized pieces per package. The Safe Lid has the same sieve configuration as the standard perforated lid with 300 holes of 0.6 mm diameter. This ensures an identical impact velocity of 19.6 m/s and anominal d50 of 1.1 μm. The simplified validation of the recovery rates according to EN 17141 resulted incomparable values for both perforated lids.

Catalogue number: 1.17421

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG

accessory

Product No.
Description
Pricing

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Detect microbial contamination gases in a few easy and automated steps. The MAS-100 Atmos® compressed gas sampler is simple and safe to use, and compliant with 21 CFR Part 11.

Cell culture media expertise accelerates lab-grown meat and seafood industry with animal-free formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service