Skip to Content
Merck
All Photos(3)

Key Documents

MAB427-C

Sigma-Aldrich

Anti-Cav1.1 alpha1s Antibody, clone 1A, Ascites Free

clone 1A, from mouse

Synonym(s):

Voltage-dependent L-type calcium channel subunit alpha-1S, Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, Voltage-gated calcium channel subunit alpha Cav1.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₹31,930.01

₹31,930.01


Estimated to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
₹31,930.01

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₹31,930.01


Estimated to ship on10 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A, monoclonal

species reactivity

plant, human, rabbit, mouse

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Voltage-dependent L-type calcium channel subunit alpha-1S (UniProt P07293; also known as Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle, Voltage-gated calcium channel subunit alpha Cav1.1) is encoded by the CACNA1S (also known as CACH1, CACN1, CACNL1A3) gene (Gene ID 100009585) in Oryctolagus cuniculus (rabbit) species. Voltage-dependent calcium channels (VDCC) are found in the membrane of excitable cells (e.g., muscle, glial cells, neurons) and exhibit 1000-fold greater permeability to calcium than to sodium under normal physiological conditions. There exist five types of VDCCs, including four high voltage-activated channels (Neural- or N-type channel, Residual- or R-type channel, Purkinje- or P/Q-type channel, and the dihydropyridine-sensitive Long-lasting or L-type channels) and the low voltage-activated Transient or T-type calcium channels. VDCCs exist as a complex of different subunits (α1, α2δ, β1-4, and γ), with α1 being the the ion-conducting pore-forming subunit that contains voltage-sensing machinery and the drug/toxin-binding sites. There exist ten genes coding for different α1 subunits, four of which code for L-type alpha-1 subunits (alpha-1A/Cav2.1/CACNA1A, alpha-1C/Cav1.2/CACNA1C, alpha-1D/Cav1.3/CACNA1D, and alpha-1S/Cav1.1/CACNA1S). The dihydropyridine-sensitive L-type channels are responsible for excitation-contraction coupling of skeletal, smooth, and cardiac muscle and for hormone secretion in endocrine cells. The α1S subunit (Cav1.1) contains the characteristic four homologous I–IV domains (a.a. 38-337, 418-664, 786-1068, 1105-1384) with six transmembrane α-helices each, having both its N- and C-terminal ends exposed intracellularly (a.a. 1-51, 1382-1873).

Specificity

Clone 1A targets the extracellular region of voltage-dependent L-type calcium channel subunit alpha-1S or Cav1.1 encoded by the CACNA1S gene.

Immunogen

Dihydropyridine-binding complex isolated from rabbit muscle t-tubules.
Epitope: Extracellular domain.

Application

Anti-Cav1.1 alpha1s Antibody, clone 1A, Ascites Free is an antibody against Cav1.1 alpha1s for use in Western Blotting, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Quality

Evaluated by Western Blotting in Jurkat membrane extract.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Cav1.1 alpha1s in 10 µg of Jurkat membrane extract.

Target description

~212 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service