Select a Size
About This Item
Skip To
Product Name
Anti-Polyhomeotic-like protein 2 Antibody (PHC2), from rabbit, purified by affinity chromatography
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rat, mouse
technique(s)
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Quality Level
Gene Information
human ... PHC2(1912)
1 of 4
This Item | SAB1402759 | HPA047403 | WH0001911M5 |
|---|---|---|---|
| biological source rabbit | biological source mouse | biological source rabbit | biological source mouse |
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| Gene Information human ... PHC2(1912) | Gene Information human ... PHC2(1912) | Gene Information human ... PHC2(1912) | Gene Information human ... PHC1(1911) |
| shipped in wet ice | shipped in dry ice | shipped in wet ice | shipped in dry ice |
| technique(s) immunohistochemistry: suitable, western blot: suitable | technique(s) indirect ELISA: suitable, western blot: 1-5 μg/mL | technique(s) immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:20-1:50 | technique(s) indirect ELISA: suitable, western blot: 1-5 μg/mL |
Analysis Note
Western Blotting Analysis: 1.0 µg/mL of this antibody detected Polyhomeotic-like protein 2 (PHC2) in 10 µg of rat kidney tissue lysate.
Application
Epigenetics & Nuclear Function
Nuclear Receptors
Disclaimer
General description
Immunogen
Other Notes
Physical form
Preparation Note
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


