6.18062
Silica gel
for column chromatography (60-120 mesh)
Synonym(s):
Silica gel
Select a Size
Select a Size
About This Item
Recommended Products
grade
for column chromatography (60-120 mesh)
bp
2230 °C
mp
>1600 °C
suitability
suitable for column chromatography
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- A Validated High-Performance Thin-Layer Chromatography Technique for Routine Analysis of Curcumin in Four Different Species of Curcuma Viz. C. amada, C. caesia, C. longa and C. zedoaria: This study presents a validated method for analyzing curcumin using high-performance thin-layer chromatography, which could benefit from the precision of Latch-lid™ TLC developing chambers in maintaining solvent consistency and improving detection accuracy (Gangal et al., 2023).
- Identification and quantification of glucose degradation products in heat-sterilized glucose solutions for parenteral use by thin-layer chromatography: Explores the use of TLC in identifying impurities in sterile pharmaceutical solutions, where Latch-lid™ chambers could ensure consistent analytical conditions (Leitzen et al., 2021).
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service