Skip to Content
Products
Cart
0
IN
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Silica gel
Synonym(s):
Silica gel
CAS Number:
112926-00-8
EC Number:
231-545-4
Products
Shipping
Ships Today (2)
Product Category
adsorbents (8)
desiccants (2)
Brand
Supelco (12)
Boiling Point (°C)
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
1.07735
Silica gel,
beads, desiccant ~ 2 - 5 mm
Expand
View Pricing
1.01907
Silica gel,
granules, desiccant ~ 2 - 5 mm
Expand
View Pricing
1.01905
Silica gel,
granules, desiccant ~ 0.2 - 1 mm
Expand
View Pricing
6.18418
Silica gel,
self-indicating coarse
Expand
View Pricing
288519
Silica gel,
high-purity grade, pore size 60 Å, 5-25 μm particle size, without binder, suitable for thin layer chromatography (TLC)
Expand
View Pricing
6.18609
Silica gel,
suitable for column chromatography
Expand
View Pricing
6.18062
Silica gel,
suitable for column chromatography
Expand
View Pricing
6.18608
Silica gel,
suitable for column chromatography
Expand
View Pricing
60765
Silica gel,
high-purity grade, with ~15% calcium sulfate and fluorescent indicator, GF
254
, suitable for thin layer chromatography (TLC)
Expand
View Pricing
6.17641
Silica gel,
GF 254 for TLC
Expand
View Pricing
6.17631
Silica gel,
G for TLC
Expand
View Pricing
288500
Silica gel,
high-purity grade, pore size 60 Å, 2-22 μm particle size distribution, without binder, suitable for thin layer chromatography (TLC)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Learn More