Skip to Content
Merck
All Photos(1)

Documents

422711-M

Millipore

KN-93, Water-Soluble

This KN-93 is a water-soluble version of KN-93 that acts as a reversible and competitive inhibitor of CaM kinase II (Ki = 370 nM for rat brain).

Synonym(s):

KN-93, Water-Soluble

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O4S · xH3PO4
Molecular Weight:
501.04
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 5 mg/mL

storage temp.

2-8°C

General description

Also available as a 5 mM solution in H2O(Cat. No. 508315).
A water-soluble form of the CaM kinase II inhibitor KN-93 (Cat. No. 422708. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Biochem/physiol Actions

Primary Target
Calmodulin-Dependent Protein Kinase (CaM Kinase) 2
Target Ki: 370 nM for cam kinase 2

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tombes, R.M., et al. 1995. Cell Growth Differ.6, 1063.
Chen, C., et al. 1994. Science266, 291.
Mamiya, N., et al. 1993. Biochem. Biophys. Res. Commun.195, 608.
Nicki, I., et al. 1993. Biochem. Biophys. Res. Commun.191, 255.
Sumi, M., et al. 1991. Biochem. Biophys. Res. Commun.181, 968.
Tokumitsu, H., et al. 1990. J. Biol. Chem.265, 4315.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service