Skip to Content
Merck
All Photos(1)

Documents

1.17098

Millipore

MAS-100 Iso MH® RS232 cable

for calibration of sampling heads with 10 m distance, for MAS-100 Iso NT®/ MAS-100 Iso MH® control units

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

aluminum cable(s)

Agency

non-compendial

description

RS232 interface

feature

electronic

packaging

pkg of 1 kit

cable L

10 m

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100® Iso

shipped in

ambient

storage temp.

no temp limit

General description

RS232, Kabel 10m is an additional accessory designed to be specifically used for MAS-100 ISO MH®. The RS 232 cable can be used as an extension cable between the anemometer and the air sampler control unit for calibration.

Application

MAS-100 ISO MH® RS232, Kabel 10m is an additional accessory. MAS-100 Iso family devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service