Skip to Content
Merck
All Photos(1)

Documents

1.11109

Supelco

Alkalinity Test

titrimetric, for use with MQUANT®

Synonym(s):

Alkalinity Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

analyte chemical class(es)

alkalis

Quality Level

compatibility

for use with MQUANT®

detection method

titrimetric

storage temp.

15-25°C

General description

Acid capacity to pH 8.2 and pH 4.3 Method: titrimetric with titration pipette MQuant®
The alkalinity is an important parameter for the assessment of the surface-water quality and the corrosive behaviour of water as well as for water and wastewater treatment. This MQuant® Alkalinity test (formally MColortest®) is suitable for the titrimetric determination using a graduated syringe.

Method
One sample (sample A) is titrated with hydrochloric acid against phenolphthalein until it becomes colorless (at pH 8.2), while a second sample (sample B) of the same water is titrated against a mixed indicator until its color changes to red (at pH 4.3). The values for KS8.2 and KS4.3 are determined from the corresponding consumption of titration solution.

The test measures alkalinity in e.g.
  • Groundwater and surface water, seawater
  • Drinking water and mineral water
  • Industrial water, prozess water, and wastewater
  • Boiler water
  • Aquarium water and waters from aquaculture
  • Swimming-pool water

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service