Skip to Content
Merck
All Photos(1)

Documents

1.14658

Supelco

Nitrite Test Kit, colorimetric

0.05-1.0 mg/L (NO₂⁻), MQuant®

Synonym(s):

Nitrite Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrite Test in freshwater and seawater, colorimetric, 0.05-1.0 mg/L (NO2-), for use with MCOLORTEST®

specific analyte(s)

nitrite

Quality Level

measuring range

0.05-1.0 mg/L (NO2-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 0.05 - 0.15 - 0.25 - 0.50 - 1.0 mg/l NO MQuant®
This MQuant® Nitrite test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The reagents are added to the sample, resulting in a color reaction. The concentration is determined by allocating the color to a value on a reference scale.
This test also is suitable to quickly check for nitrite interferences prior to a nitrate determination with Spectroquant® Nitrate in seawater 1.14556.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service