Skip to Content
Merck
All Photos(1)

Key Documents

857351

Sigma-Aldrich

D-Glutamic acid

≥99%

Synonym(s):

(R)-2-Aminoglutaric acid, (R)-2-Aminopentanedioic acid, D-Glu, R-(−)-1-Aminopropane-1,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein:
1723800
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99%

optical activity

[α]25/D −31.5°, c = 1 in 6 M HCl

mp

200-202 °C (subl.) (lit.)

SMILES string

N[C@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m1/s1

InChI key

WHUUTDBJXJRKMK-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Unnatural isomer of glutamic acid.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Volgraf et al.
Nature chemical biology, 2(1), 47-52 (2006-01-13)
The precise regulation of protein activity is fundamental to life. The allosteric control of an active site by a remote regulatory binding site is a mechanism of regulation found across protein classes, from enzymes to motors to signaling proteins. We
H Persson et al.
Molecular and cellular biology, 10(9), 4701-4711 (1990-09-01)
The gene encoding glutamic acid decarboxylase (GAD), the key enzyme in the synthesis of the inhibitory neurotransmitter gamma-aminobutyric acid, is shown to be expressed in the testis of several different species. Nucleotide sequence analysis of a cDNA clone isolated from
Pierre Maechler et al.
Diabetes, 51 Suppl 1, S99-102 (2002-01-30)
It is intriguing that the kinetics of glucose-stimulated insulin secretion from the in situ perfused pancreas differ between the rat and the mouse. Here we confirm that insulin release in the rat is clearly biphasic, whereas in the mouse glucose
Jia Luo et al.
Science (New York, N.Y.), 298(5592), 425-429 (2002-10-12)
The motor abnormalities of Parkinson's disease (PD) are caused by alterations in basal ganglia network activity, including disinhibition of the subthalamic nucleus (STN), and excessive activity of the major output nuclei. Using adeno-associated viral vector-mediated somatic cell gene transfer, we
Katarzyna Rogoz et al.
Molecular pharmacology, 85(2), 322-334 (2013-11-28)
The transient receptor potential cation channel subfamily V member 1 (TRPV1) is known as a thermosensor and integrator of inflammation-induced hyperalgesia. TRPV1 is expressed in a subpopulation of primary afferent neurons that express several different neurotransmitters. The role of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service