Skip to Content
Merck
All Photos(1)

Key Documents

65400

Sigma-Aldrich

2-Methoxycinnamic acid

≥97.0%

Synonym(s):

3-(2-Methoxyphenyl)acrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
Beilstein:
2209714
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

mp

182-186 °C

functional group

carboxylic acid

SMILES string

COc1ccccc1\C=C\C(O)=O

InChI

1S/C10H10O3/c1-13-9-5-3-2-4-8(9)6-7-10(11)12/h2-7H,1H3,(H,11,12)/b7-6+

InChI key

FEGVSPGUHMGGBO-VOTSOKGWSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ninghui Ma et al.
Molecules (Basel, Switzerland), 23(9) (2018-09-12)
To rapidly clarify and quantify the chemical profiling of Cinnamomi cortex a reliable and feasible strategy of chromatographic fingerprinting with a suite of chemometrics methods was developed and validated by ultra-high performance liquid chromatography coupled with diode array detection. Furthermore
Sirichai Adisakwattana et al.
International journal of molecular sciences, 13(2), 1778-1789 (2012-03-13)
Cinnamic acid and its derivatives have shown a variety of pharmacologic properties. However, little is known about the antiglycation properties of cinnamic acid and its derivatives. The present study sought to characterize the protein glycation inhibitory activity of cinnamic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service