Skip to Content
Merck
All Photos(1)

Key Documents

438863

Sigma-Aldrich

1-Phenoxy-2-propanol

93%

Synonym(s):

Propylene glycol phenyl ether, Propylene phenoxytol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
152.19
Beilstein:
1941356
MDL number:
Pricing and availability is not currently available.

Assay

93%

impurities

<7% dipropylene glycol phenyl ether

refractive index

n20/D 1.523 (lit.)

bp

243 °C (lit.)

density

1.064 g/mL at 20 °C (lit.)

SMILES string

CC(O)COc1ccccc1

InChI

1S/C9H12O2/c1-8(10)7-11-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3

InChI key

IBLKWZIFZMJLFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Weidhaas et al.
The Science of the total environment, 574, 1396-1404 (2016-08-20)
There are an estimated 30,000 chemicals in commerce used in quantities >1016kg per year in the US. Unfortunately there is limited information on the chemicals partitioning and reactivity properties. These orphaned or understudied chemicals are viewed as non-hazardous but can
Oscar Díaz-Santana et al.
Journal of chromatography. A, 1515, 187-195 (2017-08-16)
An extraction and determination method is shown for the analysis of dyes and solvents present in two types of ballpoint pen inks that are deposited onto paper. Ink extracts are analysed using a combination of gas chromatography with mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service