Merck
  • A case study for orphaned chemicals: 4-methylcyclohexanemethanol (MCHM) and propylene glycol phenyl ether (PPH) in riverine sediment and water treatment processes.
A case study for orphaned chemicals: 4-methylcyclohexanemethanol (MCHM) and propylene glycol phenyl ether (PPH) in riverine sediment and water treatment processes.
The Science of the total environment (2016-08-20)
Jennifer Weidhaas, Lian-Shin Lin, Karen Buzby
ABSTRACT
There are an estimated 30,000 chemicals in commerce used in quantities >1016kg per year in the US. Unfortunately there is limited information on the chemicals partitioning and reactivity properties. These orphaned or understudied chemicals are viewed as non-hazardous but can still pose serious economic, health, environmental and societal impacts as evidenced by the January 2014 spill of 37,900L of crude-MCHM (primarily 4-methylcyclohexanemethanol) and stripped-PPH (primarily dipropylene glycol phenyl ether and propylene glycol phenyl ether) into the Elk River near Charleston, WV. Using the Elk River spill as a case study of orphaned or understudied chemicals, experiments were undertaken to evaluate the adequacy of standard approaches to emergency drinking water treatment (oxidation and sorption to activated carbons). Further available sorption models for estimating the potential of these compounds to sorb to Elk River sediments and to activated carbons in drinking water systems were investigated. The results showed that powdered activated carbon (PAC) was the most effective sorbent. The trans-MCHM isomer was found to preferentially sorb compared to cis-MCHM. For MCHM concentrations ranging from 2 to 5mgL