Skip to Content
Merck
All Photos(1)

Key Documents

HPA019497

Sigma-Aldrich

Anti-OBSCN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Obscurin, Anti-Obscurin-MLCK, Anti-Obscurin-RhoGEF, Anti-Obscurin-myosin light chain kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RVDLTSTDYDTAADATESSSYFSAQGYLSSREQEGTESTTDEGQLPQVVEELRDLQVAPGTRLAKFQLKVKGYPAPRLYWFKDGQPLTASAHIRMTDKKILHTL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... OBSCN(84033)

General description

OBSCN (obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF) is a multidomain, sarcomeric Rho guanine nucleotide exchange factor protein belonging to the giant modular signaling proteins such as titin and nebulin. It has molecular mass of ~800kDa. It is expressed in the skeletal and cardiac muscle. It is located to chromosome 1q42 with >67 intracellular immunoglobulin (Ig) or fibronectin-3-like domains with multiple slice variants. It is composed of calmodulin-binding IQ motif, and a rho guanine nucleotide exchange factor domain in the COOH-terminal region.

Immunogen

Obscurin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

OBSCN (obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF) is majorly involved in the Ca(2+)/calmodulin, and G protein-coupled signal transduction pathway. It plays a vital role in the myofibrillogenesis and hypertrophic growth. It helps in the formation and spatial arrangements of contractile proteins for the structural integration and stabilization of myofibrils during sarcomere assembly.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74600.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Young et al.
The Journal of cell biology, 154(1), 123-136 (2001-07-13)
Vertebrate-striated muscle is assumed to owe its remarkable order to the molecular ruler functions of the giant modular signaling proteins, titin and nebulin. It was believed that these two proteins represented unique results of protein evolution in vertebrate muscle. In
Andrei B Borisov et al.
Histochemistry and cell biology, 125(3), 227-238 (2005-10-06)
Obscurin is a recently identified giant multidomain muscle protein (approximately 800 kDa) whose structural and regulatory functions remain to be defined. The goal of this study was to examine the effect of obscurin gene silencing induced by RNA interference on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service