Skip to Content
Merck
All Photos(1)

Key Documents

77614

Sigma-Aldrich

Phenol

purum, ≥96.0% (calc. on dry substance, T)

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

grade

purum

Assay

≥96.0% (calc. on dry substance, T)

form

liquid

autoignition temp.

1319 °F

expl. lim.

8.6 %

impurities

~10% water

color

faintly yellow

refractive index

n20/D 1.53

bp

182 °C (lit.)

mp

40-42 °C (lit.)

solubility

water: miscible

density

1.071 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 812-812 (1994)
Tyman JHP.
Synthetic and Natural Phenols., 270-270 (1996)
Tyman JHP.
Synthetic and Natural Phenols., 67-67 (1996)
Tyman JHP.
Synthetic and Natural Phenols., 1-1 (1996)
PREPARATION OF TRANSFORMING DEOXYRIBONUCLEIC ACID BY PHENOL TREATMENT.
H SAITO et al.
Biochimica et biophysica acta, 72, 619-629 (1963-08-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service