Skip to Content
Merck
All Photos(2)

Key Documents

MABN528

Sigma-Aldrich

Anti-Glypican-4 Antibody, clone 1C7.2

culture supernatant, clone 1C7.2, from mouse

Synonym(s):

Glypican-4, K-glypican, Secreted glypican-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₪2,237.00

₪2,237.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
₪2,237.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₪2,237.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

1C7.2, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GPC4(2239)

General description

Glypican-4 is a glycosylphosphatidylinositol (GPI)-anchored cell surface heparan sulfate proteoglycan (HSPG) that is encoded by the GPC4 gene in human. Glypican-4 belongs to the family of glypican-related integral membrane proteoglycans (GRIPs) that may be involved in the control of cell division and growth regulation. Deletion of the entire GPC4 gene and the two exons at the 3′ end of the adjacent GPC3 gene has been reported in a patient diagnosed with Simpson-Golabi-Behmel syndrome (SGBS).

Immunogen

GST-tagged recombinant protein corresponding to human Glypican-4.

Application

Anti-Glypican-4 Antibody, clone 1C7.2 is an antibody against Glypican-4 for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Glypican-4 in 10 µg of mouse whole embryo, mouse E16 brain, and human kidney tissue lysate.

Quality

Evaluated by Western Blotting in mouse E17 spinal cord tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Glypican-4 in 10 µg of mouse E17 spinal cord tissue lysate.

Target description

~62 kDa observed

Physical form

Mouse monoclonal IgG2bκ in tissue culture supernatant containing 0.05% sodium azide
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service