Skip to Content
Merck
All Photos(1)

Key Documents

MT51302662

Mettler-Toledo SevenGo Duo pH/DO

SG98 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61
Product MT51302662 is not currently sold in your country. Contact Technical Service

description

Calibration pH: Max. 5 points, 7 predefined buffer groups

DO: 1 or 2 points

Temperature range °C pH: ATC: -5.0 ... 105.0; MTC: -30 ... 130, DO: 0.0 ... 50.0 (+/- 0.2)

manufacturer/tradename

Mettler-Toledo 51302662

mV measuring range

-1999.9-1999.9, accuracy: ±0.1

battery power

4 × 1.5 V AA (NiMH accumulator 1.3 V)

pH range

-2-19.999

Looking for similar products? Visit Product Comparison Guide

General description

Includes meter, InLab® Expert Pro-ISM and InLab® OptiOx (both IP67, 1.8 m cable)
Mettler Toledo SevenGo Duo pro combines maximum measurement accuracy and an excellent range of functions with user-friendly operation and exceptional safety. SevenGo Duo pro meters are essential for special analyses, particularly in environmental, chemical and food & beverage industry. The kit versions are also well suited for outdoor use because both the meter and the sensors are watertight (IP67).
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.

The OptiOx optical measuring system for determining dissolved oxygen is based on the reliable RDO technology. Thanks to RDO (Rugged Dissolved Oxygen), measuring dissolved oxygen is easier than ever before.

Features and Benefits - SevenGo Duo pro pH/Ion/Oxygen Meter (IP67) for the Strictest Demands

Portable dual-channel meter for the parallel determination of pH, mV, rel. mV or ion concentrations and dissolved oxygen (DO)

Dissolved oxygen mode is based on optical RDO technology

Either linear or segmented 5-point pH calibration with selection from 7 predefined and one user-defined buffer groups

Manual or automatic atmospheric pressure compensation using built-in barometer

Modern data management for 500 GLP data records with date/time, sensor ID and serial number, user and sample ID

Contactless infrared interface for data transmission to a printer or PC

Intuitive, user-friendly operation using menu guidance in 10 languages and backlit display

Rugged watertight housing (IP67)

Revolutionary intelligent sensor management (ISM)

Other Notes

DO range 0.0 ... 500%, 0.00 ... 50.00 mg/L
DO accuracy ±0.1 mg/L from 0 ... 8 mg/L
±0.2 mg/L from 8 ... 20 mg/L
±10% from 20 ... 50 mg/L

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenGo is a trademark of Mettler-Toledo, Inc.
SevenGo Duo pro is a trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service