Key Documents
OH30073974
Ohaus® Pen pH Meter
Model ST10C-C
Select a Size
1 EACH
₪503.00
₪503.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
₪503.00
About This Item
UNSPSC Code:
41115603
₪503.00
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30073974
conductivity measuring range
0.00-19.99 mS/cm
battery power
4 × 1.5 V, 200 hr life
General description
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Application
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Features and Benefits
Easily replaceable electrodes, automatic shut off feature
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)