Skip to Content
Merck
All Photos(1)

Documents

PHL80506

Nitidine chloride

phyproof® Reference Substance

Synonym(s):

2,3-Dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridinium chloride, NSC 146397

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18NO4 · Cl
CAS Number:
Molecular Weight:
383.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

C[N+]1=CC2=CC(OC)=C(OC)C=C2C3=CC=C4C=C5C(OCO5)=CC4=C31.[Cl-]

InChI

1S/C21H18NO4.ClH/c1-22-10-13-7-17(23-2)18(24-3)8-15(13)14-5-4-12-6-19-20(26-11-25-19)9-16(12)21(14)22;/h4-10H,11H2,1-3H3;1H/q+1;/p-1

InChI key

QLDAACVSUMUMOR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Nitidine chloride is a natural product with anti-cancer activity. Its mechanism of action likely involves several pathways. Nitidine chloride has been found to inhibit topoisomerase I and topoisomerase II, induce cell apoptosis by activation of the caspase-dependent pathway, suppress c-Src/FAK associated signaling pathways and suppress Janus kinase 2/STAT3 signaling and the expression of STAT3-dependent target genes, including cyclin D1, Bcl-xL, and VEGF. Nitidine chloride has also been found to have anti-malaria activity.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service