Documents
Z354066
Sigma-Aldrich
BenchGuard-Extra bench liner sheets
size 20 in. (49 cm) × 24 in. (60 cm)
Synonym(s):
Bench Liner Sheets, BenchGuard Liner Sheets, BenchGuard Sheets
About This Item
UNSPSC Code:
41122300
NACRES:
NB.35
size
20 in. (49 cm) × 24 in. (60 cm)
General description
Extra absorbent for heavy spills, up to 800 mL/m2. PE underside prevents leakthrough. Convenient pre-cut benchtop-size sheets, 20 × 24 in. (49 × 60 cm).