Skip to Content
Merck
All Photos(3)

Documents

BAF246750020

SP Bel-Art® Labmat Bench Liner Sheets

Labmat sheet, white, L × W 20 in. × 24 in. , 50.38 cm x 61 cm

Synonym(s):

Scienceware® bench liners

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

packaging

pack of 100 ea

manufacturer/tradename

Bel-Art F24675-0020

L × W

20 in. × 24 in. , 50.38 cm x 61 cm

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Disposable Labmat Bench Liner speeds clean-up and safeguards work surfaces from stains, spills and wear. Simply roll up and discard after contamination.
Disposable bench and drawer liners protect surfaces from stains and wear. Polyethylene film backing is non-skid and prevents soak-through of most liquids while the absorbent upper surface traps aqueous, alcoholic and corrosive solutions.

Features and Benefits

  • Highly absorbent upper surface traps up to 420ml/m² of water, dyes, corrosive chemicals or radioactive solutions
  • Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
  • Available in 50.8cm (20”) wide by 15.2 m (50 ft.) rolls, or 50.8 x 61cm (20 x 24”) pre-cut sheets
  • Rolls are easily cut with the Science ware Safety Cutter (F21012-0000) or scissors while roll is retained in dispenser
  • Highly absorbent upper surface traps up to 420ml/m² of water, dyes, corrosive chemicals or radioactive solutions
  • Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
  • Available in 50.8cm (20”) wide by 15.2 m (50 ft.) rolls, or 50.8 x 61cm (20 x 24”) pre-cut sheets
  • Rolls are easily cut with the Scienceware Safety Cutter (F21012-0000) or scissors while roll is retained in dispenser

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Labmat is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service