S3375
Sedoheptulose anhydride monohydrate
Synonym(s):
2,7-Anhydro-β-D-altro-heptulopyranose, Sedoheptulosan
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H12O6 · H2O
CAS Number:
Molecular Weight:
210.18
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
biological source
Sedum spectabile
form
powder
optical activity
[α]/D -133 to -130 °, c = 2% (w/v) in water
SMILES string
O.[H][C@]12CO[C@](CO)(O1)[C@@H](O)[C@H](O)[C@@H]2O
InChI
1S/C7H12O6.H2O/c8-2-7-6(11)5(10)4(9)3(13-7)1-12-7;/h3-6,8-11H,1-2H2;1H2/t3-,4-,5-,6+,7-;/m1./s1
InChI key
TXADMKCMMLBFTL-CQJQEINESA-N
Looking for similar products? Visit Product Comparison Guide
Preparation Note
Prepared by a modified procedure of LaForge, F.B. and Hudson, C.S., J. Biol. Chem., 30, 61 (1917).
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dauren Biyashev et al.
Annals of the New York Academy of Sciences, 1480(1), 170-182 (2020-09-07)
Injury of the skin from exposure to toxic chemicals leads to the release of inflammatory mediators and the recruitment of immune cells. Nitrogen mustard (NM) and other alkylating agents cause severe cutaneous damage for which there are limited treatment options.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service