Skip to Content
Merck
All Photos(1)

Documents

OK

91021202, opossum kidney, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

OK, 91021202

biological source

opossum kidney

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Insulin, serotonin, alpha 2-adrenergic parathy. Hormone

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

American Opossum, kidney

Cell Line Description

Derived from proximal tubule of a kidney taken from an adult female opossum. Cells retain proximal tubule function. Possesses the following receptors: α 2 - adrenergic, insulin, serotonin, parathyroid hormone and atrial natriuretic factor.

Application

Kidney function studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service