Skip to Content
Merck
All Photos(1)

Documents

KG1a

91030101, human bone marrow, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KG1a, 91030101

biological source

human bone marrow

growth mode

Suspension

karyotype

2n = 46, pseudodiploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian bone marrow acute myelogenous leukaemia

Cell Line Description

Derived from KG-1, however KG1a is unresponsive to colony stimulating factor in soft agar culture and does not express the Ia-like antigen. KG1a is resistant to phorbol-diester induced macrophage differentiation and cell proliferation. KG1a cells are morphologically, cytochemically and functionally less mature than the parental KG-1. Expresses HLA A30,31; B35; CW4.

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 7
D13S317: 11,12
D16S539: 10,11
D5S818: 13
D7S820: 8,10
THO1: 7,8
TPOX: 7,9
vWA: 14,19

Culture Medium

IDMEM + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 5x100,000 to 1,000,000 cells/ml. Do not culture below 100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service