Skip to Content
Merck
All Photos(1)

Documents

A6236

Sigma-Aldrich

Agarose - TAE Blend 1.0%

Synonym(s):

Agarose - TAE Blend

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

powder

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Agarose - TAE powder blends are a TAE buffer already mixed in with agarose at a 1X concentration. To use just add water, heat and pour the gel.

Preparation Note

Preparation Instructions
1. Weigh out 1.632 g of Agarose-TAE blend 1.0% into a beaker or flask.
2. Add 100 mL of deionized water.
3. Microwave or autoclave according to method of choice for agarose gel preparation.
4. Pour the gel.
5. Perform electrophoresis according to standard TAE procedures.
6. Stain gel according to standard practices and analyze results.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service