Skip to Content
Merck
All Photos(1)

Documents

Z742590

Accuris SmartDoc 2.0 Imaging Enclosure

for use with UV or visible blue light transilluminators.

Synonym(s):

transilluminator enclosure

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000

feature

for use with UV or visible blue light transilluminators.

packaging

pkg of 1

manufacturer/tradename

Benchmark Scientific E5001-SD

General description

The SmartDoc Imaging Enclosure can be used on most existing UV or visible blue light transilluminators.

An orange photo filter is included for photography of the gels when using a visible blue light transilluminator for stain excitation. For imaging gels on a UV transilluminator, there are 2 optional photo filters available: 535nm filter for imaging green stains on UV light, or 590nm filter for imaging EtBr on UV light.

An extending adapter is included with the enclosure and can be inserted for full dimension imaging of gels up to 15 x 15 cm. Or remove the adapter for close up pictures or smaller gels.

When using the SmartDoc enclosure on transilluminators with a large illuminating surface (exceeding 19x19cm), use E5000-MAT to block light around the edge

Legal Information

Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service