Skip to Content
Merck
All Photos(1)

Documents

MABN1602

Sigma-Aldrich

Anti-PIT-1 (POU1F1) Antibody, clone 8B6.1

clone 8B6.1, from mouse

Synonym(s):

Pituitary-specific positive transcription factor 1, GHF-1, Growth hormone factor 1, PIT-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8B6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... POU1F1 (5449)

General description

Pituitary-specific positive transcription factor 1 (UniProt P28069; also known as GHF-1, Growth hormone factor 1, PIT-1) is encoded by the POU1F1 (also known as CPHD1, GHF1, PIT1) gene (Gene ID 5449) in human. PIT-1 is a transcription factor that serves as a master regulator of lineage commitment and expansion during the anterior pituitary development process that drives regional expansion and functional differentiation of five distinct hormone-producing cell lineages, including somatotropes, lactotropes, thyrotropes, corticotropes, and gonadotropes. Loss of Pit-1 expression results in a combined pituitary hormone deficiency (CPHD) syndrome reflecting the absence of lactotropes, somatotropes, and a subset of thyrotropes. Although originally characterized as pituitary-specific, PIT-1 is also expressed in human breast. PIT-1 upregulation in breast tumors increases cell proliferation and the expression of two breast cancer-related hormones, growth hormone (GH) and prolactin (PRL), leading to MMPs upregulation and breast cancer metastasis. Human POU1F1 gene encodes two alternatively spliced isoforms, where Pit-1A (GHF-2) a.a. 47-73 is truncated to a single Thr47 residue in Pit-1B (GHF-1). Both PIT-1 isoforms contain a PIT-1 POU-specific domain (a.a. 150-224 of Pit-1A or a.a. 124-198 of Pit-1B) and a DNA-binding Homeobox (a.a. 240-299 of Pit-1A or a.a. 214-273 of Pit-1B).

Specificity

Clone 8B6.1 epitope lies within an internal region present in both human PIT-1 spliced isoform A (Pit-1A, GHF-2) and isoform B (Pit-1B, GHF-1).

Immunogen

GST-tagged recombinant human PIT-1 internal fragment.

Application

Anti-PIT-1 (POU1F1), clone 8B6.1 Antibody, Cat. No. MABN1602, is a highly specific mouse monoclonal antibody, that targets POU1F1 and has been tested in Western Blotting.

Quality

Evaluated by Western Blotting in human pituitary tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PIT-1/POU1F1 in 10 µg of human pituitary tissue lysate.

Target description

~33 kDa observed. 35.67 kDa (isoform A, Pit-1A, GHF-2) and 32.91 kDa (isoform B, Pit-1B, GHF-1) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service